Gene Transcripts

Gene Symbol
SUCLG2
Entrez Gene ID
8801
Gene Name
succinate-CoA ligase GDP-forming beta subunit
Gene Aliases
GBETA
Chromosome Location
Chr. 3: 67240957 - 67654614 on Build GRCh38
UniGene ID
Hs.644919
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177599.1 NP_001171070.1 9 1056
NM_003848.3 NP_003839.2 9 1056
XM_017007415.1 XP_016862904.1 9 1052
XM_017007416.1 XP_016862905.1 9 1052
XM_017007417.1 XP_016862906.1 9 1052
XM_017007418.1 XP_016862907.1 9 1052
XM_017007419.1 XP_016862908.1 9 1052
XM_017007420.1 XP_016862909.1 9 1052
GenBank mRNA AF058954.1 - 8 944
AF131748.1 - 3 329
AK294598.1 - 9 1049
AK310527.1 - 9 1052
BC007716.2 - 9 1019 4303307
BC019868.1 - 6 695
BC035149.1 - 8 818
BC047024.1 - 10 1026 5415743
BC068602.1 - 9 1057 5274341

View Full Product Details