Gene Transcripts

Gene Symbol
PEX16
Entrez Gene ID
9409
Gene Name
peroxisomal biogenesis factor 16
Gene Aliases
PBD8A, PBD8B
Chromosome Location
Chr. 11: 45909669 - 45918123 on Build GRCh38
UniGene ID
Hs.100915
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF118240.1 - 9 1113
BC000467.2 - 9 887 2822827
BC004356.1 - 9 879 2822311

View Full Product Details