Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_197975.2 | NP_932079.1 | 4 | 883 | |
GenBank mRNA | AB020625.1 | - | 4 | 647 | |
AK025267.1 | - | 4 | 861 | ||
AY358385.1 | - | 4 | 832 | ||
BC100800.1 | - | 4 | 769 | 40000697 | |
BC100801.1 | - | 4 | 769 | 40000698 | |
BC100802.1 | - | 4 | 769 | 40000699 |