Gene Transcripts

Gene Symbol
COPS8
Entrez Gene ID
10920
Gene Name
COP9 signalosome subunit 8
Gene Aliases
COP9, CSN8, SGN8
Chromosome Location
Chr. 2: 237085441 - 237098846 on Build GRCh38
UniGene ID
Hs.531713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006710.4 NP_006701.1 2, 3 672
NM_198189.2 NP_937832.1 3, 4 737
GenBank mRNA AI690743.1 - 2, 3 401
AK289891.1 - 3, 4 271
AK294078.1 - 2, 3 227
AK308962.1 - 2, 3 199
AK313437.1 - 2, 3 215
BC003090.1 - 2, 3 196 3503287
BC036499.1 - 2, 3 806 5264527
BC080617.1 - 2, 3 168 6252628
BF979628.1 - 301
BG721393.1 - 3, 4 275
CR456994.1 - 2, 3 148
U51205.1 - 2, 3 197

View Full Product Details