Gene Transcripts

Gene Symbol
OTP
Entrez Gene ID
23440
Gene Name
orthopedia homeobox
Gene Aliases
Chromosome Location
Chr. 5: 77628712 - 77638697 on Build GRCh38
UniGene ID
Hs.202247
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032109.2 NP_115485.1 2 495
GenBank mRNA BC001800.1 - 2 495

View Full Product Details