Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015656.1 | NP_056471.1 | 13, 14 | 5324 | |
XM_011536641.2 | XP_011534943.1 | 10, 11 | 4469 | ||
GenBank mRNA | AB033062.2 | - | 12, 13 | 5198 | |
AB290161.1 | - | 12, 13 | 5294 | ||
AK026406.1 | - | 2, 3 | 374 | ||
AL117589.1 | - | 2, 3 | 484 | ||
BC009415.1 | - | 2, 3 | 517 | ||
BC025353.1 | - | 2, 3 | 1475 |