Gene Transcripts

Gene Symbol
FBXO10
Entrez Gene ID
26267
Gene Name
F-box protein 10
Gene Aliases
FBX10, PRMT11
Chromosome Location
Chr. 9: 37509920 - 37576459 on Build GRCh38
UniGene ID
Hs.709527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012166.2 NP_036298.2 2 618
XM_005251439.4 XP_005251496.1 2 707
XM_017014618.1 XP_016870107.1 2 824
XM_017014619.1 XP_016870108.1 2 935
GenBank mRNA AB209610.1 - 1 206
AF176705.1 - 2 617
BC125125.1 - 2 697 40119019
BC140785.1 - 1 589 9021653
BC171785.1 - 1 589 9054439

View Full Product Details