Gene Transcripts
- Gene Symbol
- TTTY1
- Entrez Gene ID
- 50858
- Gene Name
- testis-specific transcript, Y-linked 1 (non-protein coding)
- Gene Aliases
- LINC00033, NCRNA00033, TTY1
- Chromosome Location
- Chr. Y: 9753156 - 9774319 on Build GRCh38
- UniGene ID
- Hs.722842
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001538.2 |
|
1 |
169 |
|
GenBank mRNA |
DB035198.1 |
- |
1 |
169 |
|
- Gene Symbol
- TTTY1B
- Entrez Gene ID
- 100101116
- Gene Name
- testis-specific transcript, Y-linked 1B (non-protein coding)
- Gene Aliases
- TTTY1
- Chromosome Location
- Chr. Y: 6390401 - 6411564 on Build GRCh38
- UniGene ID
- Hs.722842
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003589.1 |
|
1 |
169 |
|
View Full Product Details