Gene Transcripts

Gene Symbol
TTTY1
Entrez Gene ID
50858
Gene Name
testis-specific transcript, Y-linked 1 (non-protein coding)
Gene Aliases
LINC00033, NCRNA00033, TTY1
Chromosome Location
Chr. Y: 9753156 - 9774319 on Build GRCh38
UniGene ID
Hs.722842
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001538.2 4 595

Gene Symbol
TTTY1B
Entrez Gene ID
100101116
Gene Name
testis-specific transcript, Y-linked 1B (non-protein coding)
Gene Aliases
TTTY1
Chromosome Location
Chr. Y: 6390401 - 6411564 on Build GRCh38
UniGene ID
Hs.722842
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003589.1 4 595

View Full Product Details