Gene Transcripts

Gene Symbol
MEMO1
Entrez Gene ID
51072
Gene Name
mediator of cell motility 1
Gene Aliases
C2orf4, CGI-27, MEMO, NS5ATP7
Chromosome Location
Chr. 2: 31867188 - 32011052 on Build GRCh38
UniGene ID
Hs.444969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137602.2 NP_001131074.1 6 940
NM_001301833.1 NP_001288762.1 8 720
NM_001301852.1 NP_001288781.1 6 499
NM_015955.3 NP_057039.1 7 1009
XM_011532892.2 XP_011531194.1 8 657
XM_011532896.2 XP_011531198.1 5 392
XM_017004255.1 XP_016859744.1 8 630
GenBank mRNA AB041018.1 - 7 1009
AB041019.1 - 7 658
AF132961.1 - 7 707
AF363446.1 - 7 926
AF529368.1 - 7 592
AK290753.1 - 8 676
AK297128.1 - 6 651
BC018733.1 - 7 974 4851517
BC070046.1 - 8 749 5273070
BC094681.1 - 7 613 6205399
BG033287.1 - 6 448

View Full Product Details