Gene Transcripts
- Gene Symbol
- EPN3
- Entrez Gene ID
- 55040
- Gene Name
- epsin 3
- Gene Aliases
- Chromosome Location
- Chr. 17: 50532687 - 50543750 on Build GRCh38
- UniGene ID
- Hs.670090
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017957.2 |
NP_060427.2
|
8, 9 |
1792 |
|
GenBank mRNA |
AF324241.1 |
- |
7, 8 |
1357 |
|
AK000785.1 |
- |
8, 9 |
1741 |
|
AK291658.1 |
- |
8, 9 |
1741 |
|
AK292977.1 |
- |
8, 9 |
1741 |
|
AK295730.1 |
- |
7, 8 |
1349 |
|
AK296346.1 |
- |
8, 9 |
1578 |
|
AK300642.1 |
- |
9, 10 |
1240 |
|
BC001038.2 |
- |
6, 7 |
1511 |
|
BC051365.1 |
- |
6, 7 |
1511 |
|
BC077722.1 |
- |
6, 7 |
1511 |
|
BC110602.2 |
- |
7, 8 |
1324 |
|
BC110605.2 |
- |
7, 8 |
1530 |
40027860 |
View Full Product Details