Gene Transcripts
- Gene Symbol
- ATG16L1
- Entrez Gene ID
- 55054
- Gene Name
- autophagy related 16 like 1
- Gene Aliases
- APG16L, ATG16A, ATG16L, IBD10, WDR30
- Chromosome Location
- Chr. 2: 233251571 - 233295674 on Build GRCh38
- UniGene ID
- Hs.529322
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190266.1 |
NP_001177195.1
|
16, 17 |
1881 |
|
NM_001190267.1 |
NP_001177196.1
|
16, 17 |
1879 |
|
NM_017974.3 |
NP_060444.3
|
15, 16 |
1826 |
|
NM_030803.6 |
NP_110430.5
|
16, 17 |
1883 |
|
NM_198890.2 |
NP_942593.2
|
12, 13 |
1394 |
|
XM_005246082.2 |
XP_005246139.1
|
17, 18 |
1871 |
|
XM_005246084.2 |
XP_005246141.1
|
14, 15 |
1410 |
|
XM_005246086.2 |
XP_005246143.1
|
13, 14 |
1359 |
|
XM_006712608.2 |
XP_006712671.1
|
16, 17 |
1682 |
|
GenBank mRNA |
AK001690.1 |
- |
15, 16 |
1671 |
|
AK024453.1 |
- |
14, 15 |
2694 |
|
AK026330.1 |
- |
3, 4 |
361 |
|
AK027854.1 |
- |
14, 15 |
1408 |
|
AK074434.1 |
- |
10, 11 |
853 |
|
AK123876.1 |
- |
13, 14 |
1245 |
|
AK308400.1 |
- |
15, 16 |
1722 |
|
AL834526.1 |
- |
2, 3 |
91 |
|
AY358182.1 |
- |
15, 16 |
2045 |
|
AY398617.1 |
- |
16, 17 |
1772 |
|
BC071846.1 |
- |
12, 13 |
1232 |
|
BC117337.1 |
- |
16, 17 |
1545 |
|
BC143900.1 |
- |
15, 16 |
1488 |
|
EF079889.1 |
- |
15, 16 |
1569 |
|
EF079890.1 |
- |
12, 13 |
1137 |
|
EF560734.1 |
- |
15, 16 |
1696 |
|
JQ924062.1 |
- |
15, 16 |
1569 |
|
View Full Product Details