Gene Transcripts

Gene Symbol
KIF26B
Entrez Gene ID
55083
Gene Name
kinesin family member 26B
Gene Aliases
Chromosome Location
Chr. 1: 245154985 - 245703126 on Build GRCh38
UniGene ID
Hs.143134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018012.3 NP_060482.2 7 2060
GenBank mRNA AK293964.1 - 7 1975
AK297303.1 - 7 2056
AY923834.1 - 7 2061
BC035896.1 - 4 581

View Full Product Details