Gene Transcripts

Gene Symbol
HEATR1
Entrez Gene ID
55127
Gene Name
HEAT repeat containing 1
Gene Aliases
BAP28, UTP10
Chromosome Location
Chr. 1: 236549005 - 236604541 on Build GRCh38
UniGene ID
Hs.708114
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018072.5 NP_060542.4 36 5232
GenBank mRNA AK125663.1 - 18 2449
AK316413.1 - 16 2203
BC035302.1 - 7 807
BC130000.1 - 21 3156

View Full Product Details