Gene Transcripts

Gene Symbol
RETN
Entrez Gene ID
56729
Gene Name
resistin
Gene Aliases
ADSF, FIZZ3, RETN1, RSTN, XCP1
Chromosome Location
Chr. 19: 7669086 - 7670454 on Build GRCh38
UniGene ID
Hs.283091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193374.1 NP_001180303.1 2, 3 154
NM_020415.3 NP_065148.1 2, 3 163
GenBank mRNA AF205952.1 - 1, 2 141
AF290874.1 - 2, 3 163
AF323081.1 - 2, 3 163
AY207314.1 - 2, 3 154
AY359066.1 - 1, 2 141
BC069302.1 - 2, 3 163 7262685
BC101554.1 - 2, 3 146 8069060
BC101560.1 - 2, 3 146 8069066

View Full Product Details