Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020062.3 | NP_064446.2 | 8 | 1198 | |
GenBank mRNA | AB044777.1 | - | 7 | 1089 | |
AB044786.1 | - | 6 | 759 | ||
AB052777.1 | - | 8 | 1242 | ||
AF249267.3 | - | 8 | 1143 | ||
AK298304.1 | - | 6 | 1121 | ||
BC001402.2 | - | 7 | 969 | ||
BC017446.1 | - | 5 | 792 | ||
BC028349.2 | - | 3 | 393 | ||
BC052306.1 | - | 8 | 1243 | 6576325 |