Gene Transcripts

Gene Symbol
RSRP1
Entrez Gene ID
57035
Gene Name
arginine and serine rich protein 1
Gene Aliases
C1orf63, NPD014
Chromosome Location
Chr. 1: 25242237 - 25338254 on Build GRCh38
UniGene ID
Hs.259412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321772.1 NP_001308701.1 3 1923
NM_020317.4 NP_064713.3 3 919
NR_135143.1 3 919
NR_135144.1 3 919
NR_135777.1 3 894
NR_135778.1 3 919
NR_135780.1 3 919
NR_135781.1 3 919
NR_135782.1 3 919
NR_135783.1 3 919
NR_135784.1 3 919
NR_135785.1 3 919
NR_135786.1 3 919
NR_135787.1 3 1923
NR_135788.1 3 985
NR_135789.1 3 1923
XM_011541797.1 XP_011540099.1 3 884
GenBank mRNA AF247168.1 - 3 846
AF267856.1 - 3 845
AK027318.1 - 3 854
AK226012.1 - 3 857
AK292532.1 - 3 854
AK296798.1 - 4 650
AL576262.3 - 3 550
BC001892.1 - 3 843 3534629
BC041843.1 - 3 1834 5269181
BC065040.1 - 3 832 6147639

View Full Product Details