Gene Transcripts

Gene Symbol
AGTRAP
Entrez Gene ID
57085
Gene Name
angiotensin II receptor associated protein
Gene Aliases
ATRAP
Chromosome Location
Chr. 1: 11736085 - 11750771 on Build GRCh38
UniGene ID
Hs.464438
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040194.1 NP_001035284.1 4 406
NM_001040195.1 NP_001035285.1 3 371
NM_001040196.1 NP_001035286.1 5 503
NM_001040197.1 NP_001035287.1 5 503
NM_020350.4 NP_065083.3 4 406
XM_011541799.2 XP_011540101.2 5 420
XM_011541800.2 XP_011540102.1 5 420
XM_011541802.1 XP_011540104.1 6 517
GenBank mRNA AF165187.1 - 4 282
AF312374.1 - 4 337
AK290042.1 - 4 349
BC011946.1 - 4 312
BC017328.2 - 4 344 4546585
BE782705.1 - 456
BI117521.1 - 4 378
CA866314.1 - 3 377
CD674964.1 - 5 383

View Full Product Details