Gene Transcripts

Gene Symbol
JPH2
Entrez Gene ID
57158
Gene Name
junctophilin 2
Gene Aliases
CMH17, JP-2, JP2
Chromosome Location
Chr. 20: 44111697 - 44187578 on Build GRCh38
UniGene ID
Hs.441737
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020433.4 NP_065166.2 1 892
NM_175913.3 NP_787109.2 1 892
XM_006723833.3 XP_006723896.1 1 884
GenBank mRNA AL132999.1 - 1 407
BC029957.1 - 1 892

View Full Product Details