Gene Transcripts

Gene Symbol
TENM2
Entrez Gene ID
57451
Gene Name
teneurin transmembrane protein 2
Gene Aliases
ODZ2, TEN-M2, TNM2, ten-2
Chromosome Location
Chr. 5: 166979066 - 168264159 on Build GRCh38
UniGene ID
Hs.631957
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080428.2 NP_001073897.2 14 2713
NM_001122679.1 NP_001116151.1 17 3264
XM_005265950.2 XP_005266007.1 19 3911
XM_005265952.2 XP_005266009.1 15 2638
XM_006714897.2 XP_006714960.1 19 3911
XM_011534604.2 XP_011532906.1 18 3725
XM_017009660.1 XP_016865149.1 19 3938
XM_017009661.1 XP_016865150.1 19 3938
XM_017009662.1 XP_016865151.1 18 3728
XM_017009663.1 XP_016865152.1 16 3133
XM_017009664.1 XP_016865153.1 16 3046
XM_017009665.1 XP_016865154.1 16 2842
XM_017009666.1 XP_016865155.1 16 2842
XM_017009667.1 XP_016865156.1 16 2815
XM_017009668.1 XP_016865157.1 16 4352
XM_017009669.1 XP_016865158.1 16 4184
GenBank mRNA AB032953.2 - 8 1401
BX648178.1 - 13 2307
CR936703.1 - 14 2646

View Full Product Details