Gene Transcripts

Gene Symbol
PLXNA4
Entrez Gene ID
91584
Gene Name
plexin A4
Gene Aliases
FAYV2820, PLEXA4, PLXNA4A, PLXNA4B, PRO34003
Chromosome Location
Chr. 7: 132123332 - 132648688 on Build GRCh38
UniGene ID
Hs.511454
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020911.1 NP_065962.1 14 3037
XM_005250686.4 XP_005250743.1 14 3435
XM_006716171.3 XP_006716234.1 15 3528
XM_011516676.2 XP_011514978.1 14 2943
XM_017012779.1 XP_016868268.1 13 2742
GenBank mRNA AB046770.3 - 12 1514

View Full Product Details