Gene Transcripts
- Gene Symbol
- YIPF2
- Entrez Gene ID
- 78992
- Gene Name
- Yip1 domain family member 2
- Gene Aliases
- FinGER2
- Chromosome Location
- Chr. 19: 10922185 - 10929012 on Build GRCh38
- UniGene ID
- Hs.164026
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321439.1 |
NP_001308368.1
|
2, 3 |
206 |
|
NM_001321440.1 |
NP_001308369.1
|
2, 3 |
206 |
|
NM_024029.4 |
NP_076934.1
|
2, 3 |
477 |
|
XM_011528270.2 |
XP_011526572.1
|
2, 3 |
206 |
|
XM_011528271.2 |
XP_011526573.1
|
2, 3 |
154 |
|
GenBank mRNA |
BC000056.2 |
- |
2, 3 |
125 |
|
BC013014.2 |
- |
2, 3 |
146 |
2960233 |
BM554546.1 |
- |
2, 3 |
124 |
|
HY015326.1 |
- |
2, 3 |
477 |
|
HY020827.1 |
- |
2, 3 |
211 |
|
View Full Product Details