Gene Transcripts

Gene Symbol
CARD14
Entrez Gene ID
79092
Gene Name
caspase recruitment domain family member 14
Gene Aliases
BIMP2, CARMA2, PRP, PSORS2, PSS1
Chromosome Location
Chr. 17: 80169992 - 80209331 on Build GRCh38
UniGene ID
Hs.675480
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001257970.1 NP_001244899.1 5, 6 1005
NM_024110.4 NP_077015.2 5, 6 1005
NM_052819.2 NP_438170.1 2, 3 286
NR_047566.1 6, 7 1070
XM_011525212.1 XP_011523514.1 7, 8 1526
XM_011525213.1 XP_011523515.1 7, 8 1268
XM_011525215.1 XP_011523517.1 6, 7 1328
XM_011525216.1 XP_011523518.1 6, 7 1028
XM_011525217.1 XP_011523519.1 6, 7 1084
XM_011525218.2 XP_011523520.1 6, 7 1067
XM_017025057.1 XP_016880546.1 7, 8 1526
XM_017025058.1 XP_016880547.1 7, 8 1526
GenBank mRNA AF322642.1 - 5, 6 1043
AK091123.1 - 6, 7 1028
AK296762.1 - 2, 3 226
AK309299.1 - 1, 2 316
AX746790.1 - 6, 7 1028
AY032927.1 - 4, 5 840
BC001326.1 - 2, 3 286
BC018142.1 - 5, 6 1014 3847282
EU652409.1 - 4, 5 837

View Full Product Details