Gene Transcripts

Gene Symbol
C1orf35
Entrez Gene ID
79169
Gene Name
chromosome 1 open reading frame 35
Gene Aliases
MMTAG2
Chromosome Location
Chr. 1: 228100727 - 228103462 on Build GRCh38
UniGene ID
Hs.445952
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024319.3 NP_077295.1 2, 3 483
NR_130975.1 2, 3 483
GenBank mRNA AK123377.1 - 2, 3 463
AK126087.1 - 2, 3 2432
AY137773.1 - 2, 3 344
BC002843.2 - 2, 3 342 3634226
BC062585.1 - 2, 3 339 5212995

View Full Product Details