Gene Transcripts

Gene Symbol
WDR59
Entrez Gene ID
79726
Gene Name
WD repeat domain 59
Gene Aliases
FP977
Chromosome Location
Chr. 16: 74873568 - 74985146 on Build GRCh38
UniGene ID
Hs.280951
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324171.1 NP_001311100.1 3 327
NM_001324172.1 NP_001311101.1 3 327
NM_030581.3 NP_085058.3 3 300
XM_005256146.3 XP_005256203.1 3 304
XM_011523331.2 XP_011521633.1 2 144
XM_017023667.1 XP_016879156.1 2 144
XM_017023670.1 XP_016879159.1 1 40
GenBank mRNA AK302557.1 - 3 368
BC004519.2 - 3 264 3938206
BU850134.1 - 3 252
CA777098.1 - 3 330

View Full Product Details