Gene Transcripts

Gene Symbol
CNTD2
Entrez Gene ID
79935
Gene Name
cyclin N-terminal domain containing 2
Gene Aliases
CCNP
Chromosome Location
Chr. 19: 40222208 - 40227111 on Build GRCh38
UniGene ID
Hs.631603
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024877.3 NP_079153.2 3 447
XM_006723395.3 XP_006723458.2 3 868
GenBank mRNA AK023327.1 - 2 324
AK301833.1 - 3 447

View Full Product Details