Gene Transcripts
- Gene Symbol
- KMT2A
- Entrez Gene ID
- 4297
- Gene Name
- lysine methyltransferase 2A
- Gene Aliases
- ALL-1, CXXC7, HRX, HTRX1, MLL, MLL-AF9, MLL/GAS7, MLL1, MLL1A, TET1-MLL, TRX1, WDSTS
- Chromosome Location
- Chr. 11: 118436490 - 118526832 on Build GRCh38
- UniGene ID
- Hs.258855
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021845.1 |
- |
2 |
363 |
|
BC090876.1 |
- |
1 |
836 |
|
View Full Product Details