Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308207.1 | NP_001295136.1 | 12 | 3792 | |
NM_001308208.1 | NP_001295137.1 | 11 | 3445 | ||
NM_025251.2 | NP_079527.1 | 11 | 3411 | ||
XM_011517308.1 | XP_011515610.1 | 12 | 3692 | ||
XM_011517309.1 | XP_011515611.1 | 12 | 3596 | ||
XM_011517312.2 | XP_011515614.1 | 10 | 4230 | ||
XM_017013870.1 | XP_016869359.1 | 13 | 4764 | ||
XM_017013871.1 | XP_016869360.1 | 12 | 3823 | ||
GenBank mRNA | AB051475.1 | - | 12 | 3796 | |
AK092501.1 | - | 7 | 858 | ||
AK296353.1 | - | 11 | 3442 | ||
AK303851.1 | - | 8 | 2114 | ||
BC013071.2 | - | 5 | 722 |