Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181900.2 | NP_871629.1 | 3 | 315 | |
NR_135013.1 | 2 | 265 | |||
XM_006720703.1 | XP_006720766.1 | 3 | 472 | ||
XM_017022639.1 | XP_016878128.1 | 2 | 830 | ||
GenBank mRNA | AF480304.1 | - | 3 | 231 | |
AK026352.1 | - | 2 | 223 | ||
AK097024.1 | - | 1 | 1683 | ||
AK225479.1 | - | 2 | 223 | ||
AL137657.1 | - | 2 | 265 | ||
BC004365.1 | - | 3 | 257 | 3641640 |