Gene Transcripts

Gene Symbol
TTTY9A
Entrez Gene ID
83864
Gene Name
testis-specific transcript, Y-linked 9A (non-protein coding)
Gene Aliases
NCRNA00131, TTTY9, TTY9
Chromosome Location
Chr. Y: 18729882 - 18739197 on Build GRCh38
UniGene ID
Hs.536002
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001530.1 5 1051

Gene Symbol
TTTY9B
Entrez Gene ID
425057
Gene Name
testis-specific transcript, Y-linked 9B (non-protein coding)
Gene Aliases
NCRNA00132, TTTY9, TTTY9A, TTY9
Chromosome Location
Chr. Y: 18581206 - 18590521 on Build GRCh38
UniGene ID
Hs.724282
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002159.1 5 1051

View Full Product Details