Gene Transcripts

Gene Symbol
TTTY14
Entrez Gene ID
83869
Gene Name
testis-specific transcript, Y-linked 14 (non-protein coding)
Gene Aliases
CYorf14, NCRNA00137, NCRNA00185, PRO2834, TTY14
Chromosome Location
Chr. Y: 18872501 - 19077547 on Build GRCh38
UniGene ID
Hs.138453
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001543.3 1 145
NR_125733.1 1 145
NR_125734.1 1 145
NR_125735.1 1 145
GenBank mRNA DA411985.1 - 1 145
DB155066.1 - 1 138

View Full Product Details