Gene Transcripts

Gene Symbol
ALKBH7
Entrez Gene ID
84266
Gene Name
alkB homolog 7
Gene Aliases
ABH7, SPATA11, UNQ6002
Chromosome Location
Chr. 19: 6372433 - 6375250 on Build GRCh38
UniGene ID
Hs.111099
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032306.3 NP_115682.1 2 604
XM_005259658.4 XP_005259715.1 2 180
XM_017027355.1 XP_016882844.1 2 240
GenBank mRNA AK223336.1 - 2 227
AK311956.1 - 2 243
AY358858.1 - 2 604
AY427650.2 - 2 604
BC004393.1 - 2 259 3635540
BX334356.2 - 2 521

View Full Product Details