Gene Transcripts

Gene Symbol
TRIM55
Entrez Gene ID
84675
Gene Name
tripartite motif containing 55
Gene Aliases
MURF-2, RNF29, muRF2
Chromosome Location
Chr. 8: 66112667 - 66175485 on Build GRCh38
UniGene ID
Hs.85524
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033058.2 NP_149047.2 3 676
NM_184085.1 NP_908973.1 3 676
NM_184086.1 NP_908974.1 3 676
NM_184087.1 NP_908975.1 3 676
XM_005251316.4 XP_005251373.1 5 1701
XM_011517613.2 XP_011515915.1 5 1701
XM_011517614.2 XP_011515916.1 5 1624
XM_011517615.2 XP_011515917.1 5 1701
XM_011517616.2 XP_011515918.1 5 1701
XM_011517617.2 XP_011515919.1 3 758
XM_017013908.1 XP_016869397.1 3 394
XM_017013909.1 XP_016869398.1 2 213
GenBank mRNA AJ243488.1 - 3 676
AJ243489.1 - 3 676
AJ277493.1 - 3 676
AJ291712.1 - 3 582
AJ431704.1 - 3 676
AK091310.1 - 3 826
AK091728.1 - 3 496
BC007750.2 - 3 582 4110783
BT007212.1 - 3 402

View Full Product Details