Gene Transcripts

Gene Symbol
IGHV5-78
Entrez Gene ID
28387
Gene Name
immunoglobulin heavy variable 5-78 (pseudogene)
Gene Aliases
5-78P, IGHV578, VH5
Chromosome Location
Chr. 14: 106851112 - 106851546 on Build GRCh38
UniGene ID
Hs.81221
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF085654.1 - 18
BC002792.2 - 1, 2 71 3621362
X58398.1 - 1, 2 69
X58399.1 - 1, 2 52

View Full Product Details