Gene Transcripts
- Gene Symbol
- LRRC37B
- Entrez Gene ID
- 114659
- Gene Name
- leucine rich repeat containing 37B
- Gene Aliases
- LRRC37
- Chromosome Location
- Chr. 17: 32007872 - 32053500 on Build GRCh38
- UniGene ID
- Hs.514071
- Species
- Homo sapiens
- Gene Symbol
- LRRC37A2
- Entrez Gene ID
- 474170
- Gene Name
- leucine rich repeat containing 37 member A2
- Gene Aliases
- LRRC37
- Chromosome Location
- Chr. 17: 46512657 - 46556747 on Build GRCh38
- UniGene ID
- Hs.646568
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
JQ790576.1 |
- |
8 |
715 |
|
- Gene Symbol
- LRRC37A4P
- Entrez Gene ID
- 55073
- Gene Name
- leucine rich repeat containing 37 member A4, pseudogene
- Gene Aliases
- LRRC37, LRRC37A4
- Chromosome Location
- Chr. 17: 45505883 - 45520523 on Build GRCh38
- UniGene ID
- Hs.596756
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
JQ790698.1 |
- |
8 |
715 |
|
View Full Product Details