Gene Transcripts

Gene Symbol
FHAD1
Entrez Gene ID
114827
Gene Name
forkhead associated phosphopeptide binding domain 1
Gene Aliases
Chromosome Location
Chr. 1: 15236559 - 15400283 on Build GRCh38
UniGene ID
Hs.659997
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052929.1 NP_443161.1 4 498
XM_011540575.2 XP_011538877.1 4 574
XM_011540576.2 XP_011538878.1 4 575
XM_011540577.2 XP_011538879.1 4 536
XM_011540580.2 XP_011538882.1 4 573
XM_011540581.2 XP_011538883.1 4 573
XM_011540582.2 XP_011538884.1 4 573
XM_011540584.2 XP_011538886.1 4 574
XM_011540588.2 XP_011538890.1 4 565
XM_011540589.2 XP_011538891.1 4 574
XM_011540590.2 XP_011538892.1 4 566
XM_011540592.2 XP_011538894.1 3 275
XM_011540593.2 XP_011538895.1 4 570
XM_011540597.2 XP_011538899.1 4 575
XM_011540598.2 XP_011538900.1 4 577
XM_017000198.1 XP_016855687.1 4 575
XM_017000199.1 XP_016855688.1 4 575
XM_017000200.1 XP_016855689.1 4 574
XM_017000201.1 XP_016855690.1 4 574
XM_017000202.1 XP_016855691.1 4 575
XM_017000203.1 XP_016855692.1 4 571
XM_017000204.1 XP_016855693.1 4 570
XM_017000205.1 XP_016855694.1 4 575
XM_017000206.1 XP_016855695.1 4 570
XM_017000207.1 XP_016855696.1 4 576
XM_017000208.1 XP_016855697.1 4 565
XM_017000209.1 XP_016855698.1 4 569
XM_017000210.1 XP_016855699.1 4 565
XM_017000211.1 XP_016855700.1 4 573
XM_017000212.1 XP_016855701.1 4 564
XM_017000213.1 XP_016855702.1 4 574
XM_017000214.1 XP_016855703.1 4 575
GenBank mRNA AK057963.1 - 4 499

View Full Product Details