Gene Transcripts

Gene Symbol
NXNL1
Entrez Gene ID
115861
Gene Name
nucleoredoxin-like 1
Gene Aliases
RDCVF, TXNL6
Chromosome Location
Chr. 19: 17455425 - 17460916 on Build GRCh38
UniGene ID
Hs.661836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138454.1 NP_612463.1 1 76
GenBank mRNA BC014127.1 - 1 76
DQ426894.1 - 1 114

View Full Product Details