Gene Transcripts

Gene Symbol
DRGX
Entrez Gene ID
644168
Gene Name
dorsal root ganglia homeobox
Gene Aliases
DRG11, PRRXL1
Chromosome Location
Chr. 10: 49364085 - 49396016 on Build GRCh38
UniGene ID
Hs.534530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011540089.2 XP_011538391.1 4 397

View Full Product Details