Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153340.4 | NP_699171.3 | 5 | 586 | |
XM_005270445.3 | XP_005270502.1 | 5 | 717 | ||
XM_005270446.2 | XP_005270503.1 | 5 | 713 | ||
XM_011540638.2 | XP_011538940.1 | 4 | 616 | ||
XM_011540639.2 | XP_011538941.1 | 5 | 1986 | ||
XM_011540641.2 | XP_011538943.1 | 5 | 1319 | ||
XM_011540644.2 | XP_011538946.1 | 7 | 801 | ||
XM_011540645.1 | XP_011538947.1 | 5 | 645 | ||
XM_011540646.2 | XP_011538948.1 | 5 | 710 | ||
XM_017000267.1 | XP_016855756.1 | 6 | 1448 | ||
XM_017000268.1 | XP_016855757.1 | 6 | 781 | ||
XM_017000269.1 | XP_016855758.1 | 6 | 1448 | ||
XM_017000270.1 | XP_016855759.1 | 6 | 849 | ||
GenBank mRNA | AK090460.1 | - | 3 | 1434 |