Gene Transcripts

Gene Symbol
TDH
Entrez Gene ID
157739
Gene Name
L-threonine dehydrogenase (pseudogene)
Gene Aliases
SDR14E1P
Chromosome Location
Chr. 8: 11339637 - 11368452 on Build GRCh38
UniGene ID
Hs.583896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001578.1 6 585
GenBank mRNA AJ301562.1 - 6 585
AK057762.1 - 6 524
AY101186.1 - 5 487
AY101187.1 - 4 337
BC074983.2 - 5 372

View Full Product Details