Gene Transcripts

Gene Symbol
GRASP
Entrez Gene ID
160622
Gene Name
general receptor for phosphoinositides 1 associated scaffold protein
Gene Aliases
TAMALIN
Chromosome Location
Chr. 12: 52006945 - 52015889 on Build GRCh38
UniGene ID
Hs.407202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271856.1 NP_001258785.1 4 367
NM_181711.3 NP_859062.1 5 601
XM_005268691.3 XP_005268748.1 5 1769
XM_011537996.1 XP_011536298.1 4 1547
GenBank mRNA BC035500.2 - 4 338
BC052967.1 - 4 285
BC053666.1 - 5 582 6151168
HY035281.1 - 4 301

View Full Product Details