Gene Transcripts

Gene Symbol
RGS21
Entrez Gene ID
431704
Gene Name
regulator of G-protein signaling 21
Gene Aliases
Chromosome Location
Chr. 1: 192316992 - 192367285 on Build GRCh38
UniGene ID
Hs.558673
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039152.3 NP_001034241.1 5 556
GenBank mRNA AY643711.1 - 5 557

View Full Product Details