Gene Transcripts
- Gene Symbol
- PTCRA
- Entrez Gene ID
- 171558
- Gene Name
- pre T-cell antigen receptor alpha
- Gene Aliases
- PT-ALPHA, PTA
- Chromosome Location
- Chr. 6: 42915989 - 42925838 on Build GRCh38
- UniGene ID
- Hs.169002
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243168.1 |
NP_001230097.1
|
2, 3 |
454 |
|
NM_001243169.1 |
NP_001230098.1
|
3, 4 |
379 |
|
NM_138296.2 |
NP_612153.2
|
2, 3 |
454 |
|
GenBank mRNA |
AF165312.1 |
- |
3, 4 |
298 |
|
AK130819.1 |
- |
|
388 |
|
AK310559.1 |
- |
2, 3 |
518 |
|
AK312223.1 |
- |
2, 3 |
387 |
|
BC069336.1 |
- |
2, 3 |
422 |
7262230 |
BC100771.1 |
- |
2, 3 |
396 |
40000277 |
BC100772.1 |
- |
2, 3 |
396 |
40000280 |
BC100773.2 |
- |
2, 3 |
396 |
40000282 |
BC100774.2 |
- |
2, 3 |
396 |
40000286 |
BC153829.1 |
- |
2, 3 |
396 |
40000274 |
U36759.1 |
- |
2, 3 |
454 |
|
U38996.1 |
- |
2, 3 |
339 |
|
View Full Product Details