Gene Transcripts

Gene Symbol
B3GNT6
Entrez Gene ID
192134
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Gene Aliases
B3Gn-T6, BGnT-6, beta-1,3-Gn-T6, beta3Gn-T6
Chromosome Location
Chr. 11: 77034341 - 77041973 on Build GRCh38
UniGene ID
Hs.352622
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138706.4 NP_619651.3 2 594
GenBank mRNA AB073740.1 - 1 456
AK127544.1 - 1 270
AK172863.1 - 2 519
AK292773.1 - 2 537
AK313682.1 - 2 525
BC025357.1 - 1 135
BC103908.1 - 2 493 40005856
BC103909.2 - 2 493 40005863
BC103910.1 - 2 493 40005864
DA437068.1 - 2 544

View Full Product Details