Gene Transcripts
- Gene Symbol
- SLC12A3
- Entrez Gene ID
- 6559
- Gene Name
- solute carrier family 12 member 3
- Gene Aliases
- NCC, NCCT, TSC
- Chromosome Location
- Chr. 16: 56865207 - 56915850 on Build GRCh38
- UniGene ID
- Hs.669115
- Species
- Homo sapiens
- Gene Symbol
- DOCK10
- Entrez Gene ID
- 55619
- Gene Name
- dedicator of cytokinesis 10
- Gene Aliases
- DRIP2, Nbla10300, ZIZ3
- Chromosome Location
- Chr. 2: 224765090 - 225042689 on Build GRCh38
- UniGene ID
- Hs.46578
- Species
- Homo sapiens
- Gene Symbol
- DPH3P1
- Entrez Gene ID
- 100132911
- Gene Name
- diphthamide biosynthesis 3 pseudogene 1
- Gene Aliases
- C20orf143, DPH3B, ZCSL1
- Chromosome Location
- Chr. 20: 62844566 - 62846191 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080750.4 |
NP_542788.1
|
1 |
167 |
|
- Gene Symbol
- HERC6
- Entrez Gene ID
- 55008
- Gene Name
- HECT and RLD domain containing E3 ubiquitin protein ligase family member 6
- Gene Aliases
- Chromosome Location
- Chr. 4: 88378565 - 88443097 on Build GRCh38
- UniGene ID
- Hs.529317
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL832670.1 |
- |
1 |
1727 |
|
- Gene Symbol
- PABPC1P2
- Entrez Gene ID
- 728773
- Gene Name
- poly(A) binding protein cytoplasmic 1 pseudogene 2
- Gene Aliases
- PABP2, PABP4, PABPCP2, PABPCP4
- Chromosome Location
- Chr. 2: 146587057 - 146590990 on Build GRCh38
- UniGene ID
- Hs.334462
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026904.1 |
|
1 |
3254 |
|
GenBank mRNA |
BC068242.1 |
- |
1 |
2619 |
|
- Gene Symbol
- SPTSSA
- Entrez Gene ID
- 171546
- Gene Name
- serine palmitoyltransferase small subunit A
- Gene Aliases
- C14orf147, SSSPTA
- Chromosome Location
- Chr. 14: 34432938 - 34462262 on Build GRCh38
- UniGene ID
- Hs.740577
- Species
- Homo sapiens
- Gene Symbol
- U2AF1L4
- Entrez Gene ID
- 199746
- Gene Name
- U2 small nuclear RNA auxiliary factor 1 like 4
- Gene Aliases
- U2AF1-RS3, U2AF1L3, U2AF1L3V1, U2AF1RS3, U2af26
- Chromosome Location
- Chr. 19: 35742527 - 35745442 on Build GRCh38
- UniGene ID
- Hs.351558
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092844.1 |
- |
3 |
724 |
|
AK094213.1 |
- |
3 |
1545 |
|
AK303983.1 |
- |
4 |
1373 |
|
- Gene Symbol
- ZNF749
- Entrez Gene ID
- 388567
- Gene Name
- zinc finger protein 749
- Gene Aliases
- Chromosome Location
- Chr. 19: 57428565 - 57447097 on Build GRCh38
- UniGene ID
- Hs.467358
- Species
- Homo sapiens
View Full Product Details