Gene Transcripts

Gene Symbol
CMYA5
Entrez Gene ID
202333
Gene Name
cardiomyopathy associated 5
Gene Aliases
C5orf10, SPRYD2, TRIM76
Chromosome Location
Chr. 5: 79689339 - 79800226 on Build GRCh38
UniGene ID
Hs.482625
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153610.4 NP_705838.3 2 2617
XM_017009212.1 XP_016864701.1 2 2485
GenBank mRNA AL832347.1 - 2 2419
AL832376.1 - 2 2576

View Full Product Details