Gene Transcripts
- Gene Symbol
- TTTY17A
- Entrez Gene ID
- 252949
- Gene Name
- testis-specific transcript, Y-linked 17A (non-protein coding)
- Gene Aliases
- NCRNA00140, TTTY17
- Chromosome Location
- Chr. Y: 22851584 - 22852715 on Build GRCh38
- UniGene ID
- Hs.522869
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001526.1 |
|
2 |
146 |
|
- Gene Symbol
- TTTY17B
- Entrez Gene ID
- 474151
- Gene Name
- testis-specific transcript, Y-linked 17B (non-protein coding)
- Gene Aliases
- NCRNA00141
- Chromosome Location
- Chr. Y: 24485332 - 24486463 on Build GRCh38
- UniGene ID
- Hs.741142
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002180.1 |
|
2 |
146 |
|
- Gene Symbol
- TTTY17C
- Entrez Gene ID
- 474152
- Gene Name
- testis-specific transcript, Y-linked 17C (non-protein coding)
- Gene Aliases
- NCRNA00142
- Chromosome Location
- Chr. Y: 25183643 - 25184773 on Build GRCh38
- UniGene ID
- Hs.741142
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002179.1 |
|
2 |
146 |
|
View Full Product Details