Gene Transcripts
- Gene Symbol
- TTTY23
- Entrez Gene ID
- 252955
- Gene Name
- testis-specific transcript, Y-linked 23 (non-protein coding)
- Gene Aliases
- NCRNA00148, TTTY23B
- Chromosome Location
- Chr. Y: 9910798 - 9911962 on Build GRCh38
- UniGene ID
- Hs.522846
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001540.1 |
|
3 |
302 |
|
- Gene Symbol
- TTTY23B
- Entrez Gene ID
- 100101121
- Gene Name
- testis-specific transcript, Y-linked 23B (non-protein coding)
- Gene Aliases
- TTTY23
- Chromosome Location
- Chr. Y: 6242446 - 6243610 on Build GRCh38
- UniGene ID
- Hs.522846
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003593.1 |
|
3 |
302 |
|
View Full Product Details