Gene Transcripts

Gene Symbol
STXBP4
Entrez Gene ID
252983
Gene Name
syntaxin binding protein 4
Gene Aliases
Synip
Chromosome Location
Chr. 17: 54968728 - 55164285 on Build GRCh38
UniGene ID
Hs.35199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178509.5 NP_848604.3 7, 8 769
XM_005257187.4 XP_005257244.1 7, 8 811
XM_006721797.3 XP_006721860.1 7, 8 809
XM_006721798.3 XP_006721861.1 7, 8 800
XM_017024410.1 XP_016879899.1 8, 9 860
XM_017024411.1 XP_016879900.1 8, 9 852
XM_017024412.1 XP_016879901.1 7, 8 803
XM_017024413.1 XP_016879902.1 5, 6 524
XM_017024414.1 XP_016879903.1 8, 9 859
XM_017024415.1 XP_016879904.1 8, 9 859
GenBank mRNA AK122865.1 - 7, 8 769
BC041485.2 - 6, 7 612
BC125116.1 - 7, 8 758 40118892

View Full Product Details