Gene Transcripts
- Gene Symbol
- SERHL2
- Entrez Gene ID
- 253190
- Gene Name
- serine hydrolase-like 2
- Gene Aliases
- dJ222E13.1
- Chromosome Location
- Chr. 22: 42553857 - 42574382 on Build GRCh38
- UniGene ID
- Hs.728878
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001284334.1 |
NP_001271263.1
|
6, 7 |
579 |
|
NM_014509.4 |
NP_055324.2
|
8, 9 |
771 |
|
XM_017028739.1 |
XP_016884228.1
|
7, 8 |
716 |
|
GenBank mRNA |
AL450314.1 |
- |
8, 9 |
714 |
|
AL589866.1 |
- |
2, 3 |
72 |
|
AL590118.1 |
- |
6, 7 |
420 |
|
AL590120.1 |
- |
8, 9 |
612 |
|
BC119650.2 |
- |
8, 9 |
655 |
40115836 |
BC119651.1 |
- |
8, 9 |
655 |
40115837 |
BC127635.1 |
- |
8, 9 |
656 |
40115838 |
BC127636.1 |
- |
8, 9 |
656 |
40115841 |
CR456455.1 |
- |
2, 3 |
78 |
|
View Full Product Details